siRNA Details
| Gene Symbol: ELMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024712.3 | NP_078988.2 | 9 | 984 | ||
| GenBank mRNA | AK023886.1 | - | 8 | 748 | ||
| AK056923.1 | - | 8 | 947 | |||
| AK300976.1 | - | 9 | 998 | |||
| BC034410.1 | - | 6 | 596 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ELMO3| Gene Symbol: ELMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024712.3 | NP_078988.2 | 9 | 984 | ||
| GenBank mRNA | AK023886.1 | - | 8 | 748 | ||
| AK056923.1 | - | 8 | 947 | |||
| AK300976.1 | - | 9 | 998 | |||
| BC034410.1 | - | 6 | 596 | |||