siRNA Details
| Gene Symbol: KLHL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024731.3 | NP_079007.2 | 4 | 1410 | ||
| XM_005256149.2 | XP_005256206.1 | 4 | 1624 | |||
| XM_017023674.1 | XP_016879163.1 | 4 | 1372 | |||
| XM_017023675.1 | XP_016879164.1 | 4 | 1392 | |||
| GenBank mRNA | AK022605.1 | - | 3 | 1263 | ||
| AK074056.1 | - | 1 | 339 | |||
| AK122745.1 | - | 2 | 1845 | |||
| BC033821.1 | - | 4 | 1396 | 5219021 | ||

KLHL36
Genomic Map