siRNA Details
| Gene Symbol: CLMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024734.3 | NP_079010.2 | 3 | 278 | ||
| XM_011537158.1 | XP_011535460.1 | 3 | 274 | |||
| XM_011537159.2 | XP_011535461.1 | 3 | 274 | |||
| XM_017021646.1 | XP_016877135.1 | 3 | 444 | |||
| XM_017021647.1 | XP_016877136.1 | 3 | 274 | |||
| GenBank mRNA | AB047979.1 | - | 3 | 235 | ||
| AK025226.1 | - | 3 | 221 | |||
| AK314315.1 | - | 3 | 276 | |||
| AL117526.1 | - | 3 | 130 | |||
| BC068482.1 | - | 3 | 227 | 4375469 | ||
| CB155696.1 | - | 3 | 278 | |||

CLMN
Genomic Map