siRNA Details
| Gene Symbol: RNF122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024787.3 | NP_079063.2 | 2 | 509 | ||
| XM_017013846.1 | XP_016869335.1 | 2 | 405 | |||
| GenBank mRNA | AK022588.1 | - | 2 | 506 | ||
| AU124463.1 | - | 2 | 506 | |||
| BC093884.1 | - | 2 | 127 | 7939729 | ||
| BC101573.1 | - | 2 | 127 | 8069079 | ||
| D38656.1 | - | 1 | 79 | |||
| DA610129.1 | - | 2 | 509 | |||

RNF122
Genomic Map