siRNA Details
| Gene Symbol: RNF122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024787.3 | NP_079063.2 | 3 | 623 | ||
| XM_017013846.1 | XP_016869335.1 | 3 | 519 | |||
| GenBank mRNA | BC093884.1 | - | 3 | 241 | 7939729 | |
| BC101573.1 | - | 3 | 241 | 8069079 | ||
| BF847236.1 | - | 2 | 148 | |||

RNF122
Genomic Map