siRNA Details
| Gene Symbol: SLC35E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024881.4 | NP_079157.3 | 4, 5 | 775 | ||
| GenBank mRNA | AK027699.1 | - | 4, 5 | 569 | ||
| AK027850.1 | - | 2, 3 | 186 | |||
| AK075355.1 | - | 2, 3 | 214 | |||
| BC062562.1 | - | 4, 5 | 775 | 6598965 | ||

SLC35E1
Genomic Map