siRNA Details
| Gene Symbol: LINC00472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026807.2 | 4 | 1515 | |||
| NR_121612.1 | 4 | 1566 | ||||
| NR_121613.1 | 3 | 1500 | ||||
| NR_121614.1 | 3 | 1449 | ||||
| GenBank mRNA | AK023251.1 | - | 4 | 1515 | ||
| DA622283.1 | - | 1 | 237 | |||

LINC00472
Genomic Map