siRNA Details
| Gene Symbol: DSN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145315.1 | NP_001138787.1 | 4 | 581 | ||
| NM_001145316.1 | NP_001138788.1 | 4 | 773 | |||
| NM_001145317.1 | NP_001138789.1 | 3 | 260 | |||
| NM_001145318.1 | NP_001138790.1 | 3 | 532 | |||
| NM_024918.3 | NP_079194.3 | 4 | 686 | |||
| XM_006723876.2 | XP_006723939.1 | 3 | 255 | |||
| XM_017028070.1 | XP_016883559.1 | 4 | 569 | |||
| XM_017028071.1 | XP_016883560.1 | 4 | 791 | |||
| XM_017028072.1 | XP_016883561.1 | 3 | 479 | |||
| GenBank mRNA | AK023408.1 | - | 5 | 412 | ||
| AK093031.1 | - | 4 | 681 | |||
| AK290738.1 | - | 4 | 773 | |||
| AK301671.1 | - | 3 | 465 | |||
| AK301840.1 | - | 3 | 210 | |||
| BC026011.1 | - | 3 | 786 | 4827570 | ||
| BC058899.1 | - | 4 | 539 | 6063417 | ||

DSN1
Genomic Map