siRNA Details
| Gene Symbol: FRMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122841.2 | NP_001116313.1 | 4 | 355 | ||
| NM_024919.4 | NP_079195.3 | 4 | 505 | |||
| NR_110312.1 | 5 | 883 | ||||
| XM_011536137.1 | XP_011534439.1 | 5 | 585 | |||
| XM_011536138.1 | XP_011534440.1 | 5 | 401 | |||
| XM_011536140.1 | XP_011534442.1 | 4 | 440 | |||
| XM_011536141.1 | XP_011534443.1 | 4 | 380 | |||
| XM_011536142.2 | XP_011534444.1 | 4 | 418 | |||
| XM_011536143.1 | XP_011534445.1 | 4 | 316 | |||
| XM_011536145.1 | XP_011534447.1 | 4 | 355 | |||
| XM_017011317.1 | XP_016866806.1 | 5 | 585 | |||
| GenBank mRNA | AK074110.1 | - | 5 | 442 | ||
| AK097179.1 | - | 4 | 505 | |||
| AK097579.1 | - | 4 | 355 | |||
| AK125963.1 | - | 5 | 855 | |||
| AK303194.1 | - | 3 | 159 | |||
| AK303407.1 | - | 2 | 213 | |||
| AK316478.1 | - | 3 | 159 | |||
| BC137144.1 | - | 4 | 463 | 9021141 | ||

FRMD1
Genomic Map