siRNA Details
| Gene Symbol: UBA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003335.2 | NP_003326.2 | 3 | 422 | ||
| XM_005265430.2 | XP_005265487.1 | 3 | 406 | |||
| XM_006713321.2 | XP_006713384.1 | 3 | 406 | |||
| XM_011534070.1 | XP_011532372.1 | 3 | 406 | |||
| XM_011534071.1 | XP_011532373.1 | 3 | 406 | |||
| XM_011534072.1 | XP_011532374.1 | 3 | 406 | |||
| GenBank mRNA | BC006378.2 | - | 3 | 347 | 4299266 | |
| BT007026.1 | - | 3 | 251 | |||
| L13852.1 | - | 3 | 415 | |||

UBA7
Genomic Map