siRNA Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323679.1 | NP_001310608.1 | 6 | 1650 | ||
| NM_001323680.1 | NP_001310609.1 | 4 | 1731 | |||
| NM_001323681.1 | NP_001310610.1 | 4 | 1722 | |||
| NM_001323682.1 | NP_001310611.1 | 5 | 1788 | |||
| NM_001323683.1 | NP_001310612.1 | 5 | 1782 | |||
| NM_001323684.1 | NP_001310613.1 | 5 | 1810 | |||
| NM_001323685.1 | NP_001310614.1 | 4 | 1451 | |||
| NM_024958.2 | NP_079234.1 | 4 | 1709 | |||
| NR_136649.1 | 3 | 1514 | ||||
| XM_006723630.3 | XP_006723693.1 | 4 | 2132 | |||
| XM_011529360.1 | XP_011527662.1 | 4 | 1415 | |||
| XM_011529362.2 | XP_011527664.1 | 5 | 1422 | |||
| XM_011529363.2 | XP_011527665.1 | 4 | 1400 | |||
| XM_017028073.1 | XP_016883562.1 | 4 | 1354 | |||
| XM_017028074.1 | XP_016883563.1 | 4 | 1408 | |||
| XM_017028075.1 | XP_016883564.1 | 4 | 1580 | |||
| GenBank mRNA | AK026982.1 | - | 4 | 1321 | ||
| AL136915.1 | - | 3 | 1288 | |||
| BC001963.1 | - | 3 | 1288 | 3454427 | ||

NRSN2
Genomic Map