siRNA Details
| Gene Symbol: LOC107986810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012934.1 | XP_016868423.1 | 1 | 1957 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107986810| Gene Symbol: LOC107986810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012934.1 | XP_016868423.1 | 1 | 1957 | ||