siRNA Details
| Gene Symbol: ASRGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083926.1 | NP_001077395.1 | 5 | 928 | ||
| NM_025080.3 | NP_079356.3 | 5 | 851 | |||
| XM_005274305.3 | XP_005274362.1 | 4 | 568 | |||
| XM_005274306.2 | XP_005274363.1 | 4 | 630 | |||
| XM_011545265.2 | XP_011543567.1 | 5 | 810 | |||
| GenBank mRNA | AF411076.1 | - | 5 | 660 | ||
| AK090908.1 | - | 5 | 700 | |||
| AK091894.1 | - | 5 | 782 | |||
| AK225802.1 | - | 5 | 619 | |||
| AK297158.1 | - | 4 | 559 | |||
| AK309845.1 | - | 4 | 553 | |||
| AK313069.1 | - | 5 | 711 | |||
| BC006267.2 | - | 2 | 241 | |||
| BC021295.2 | - | 5 | 649 | 3937243 | ||
| BC064963.1 | - | 4 | 541 | 6141919 | ||
| BC093070.1 | - | 5 | 753 | 30370741 | ||
| BX640832.1 | - | 1 | 232 | |||

ASRGL1
Genomic Map