siRNA Details
| Gene Symbol: KIAA0226L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286761.1 | NP_001273690.1 | 5, 6 | 1417 | ||
| NM_001286762.1 | NP_001273691.1 | 4, 5 | 1323 | |||
| NM_001286763.1 | NP_001273692.1 | 5, 6 | 1052 | |||
| NM_001286764.1 | NP_001273693.1 | 4, 5 | 936 | |||
| NM_001286765.1 | NP_001273694.1 | 4, 5 | 782 | |||
| NM_001286766.1 | NP_001273695.1 | 3, 4 | 666 | |||
| NM_025113.3 | NP_079389.2 | 5, 6 | 1439 | |||
| GenBank mRNA | AK025215.1 | - | 4, 5 | 1072 | ||
| AK093073.1 | - | 5, 6 | 1387 | |||
| AK125950.1 | - | 5, 6 | 1424 | |||
| AK293034.1 | - | 4, 5 | 688 | |||
| AK300183.1 | - | 4, 5 | 486 | |||
| AK307854.1 | - | 3, 4 | 409 | |||
| AK310264.1 | - | 6, 7 | 1426 | |||
| BC004495.1 | - | 1, 2 | 136 | |||
| BC032311.1 | - | 4, 5 | 1004 | |||
| BC043488.1 | - | 5, 6 | 785 | 5205680 | ||
| BX641126.1 | - | 3, 4 | 473 | |||

KIAA0226L
Genomic Map