siRNA Details
| Gene Symbol: RUFY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040451.2 | NP_001035541.1 | 9 | 1201 | ||
| NM_001040452.2 | NP_001035542.1 | 10 | 1012 | |||
| NM_025158.4 | NP_079434.3 | 10 | 1187 | |||
| XM_005265993.4 | XP_005266050.1 | 7 | 558 | |||
| XM_006714921.3 | XP_006714984.2 | 9 | 1131 | |||
| XM_006714922.3 | XP_006714985.1 | 7 | 991 | |||
| XM_017009891.1 | XP_016865380.1 | 9 | 946 | |||
| XM_017009892.1 | XP_016865381.1 | 8 | 984 | |||
| XM_017009893.1 | XP_016865382.1 | 7 | 918 | |||
| XM_017009894.1 | XP_016865383.1 | 11 | 1153 | |||
| XM_017009895.1 | XP_016865384.1 | 10 | 1137 | |||
| GenBank mRNA | AB209507.1 | - | 11 | 1057 | ||
| AF312367.1 | - | 10 | 1178 | |||
| AF361055.2 | - | 10 | 1160 | |||
| AK025904.1 | - | 4 | 250 | |||
| AK075021.1 | - | 9 | 1101 | |||
| AK093848.1 | - | 2 | 1119 | |||
| AK291926.1 | - | 9 | 1201 | |||
| BC032571.1 | - | 10 | 931 | 5243223 | ||

RUFY1
Genomic Map