siRNA Details
| Gene Symbol: ADAM33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282447.2 | NP_001269376.1 | 22 | 3920 | ||
| NM_025220.4 | NP_079496.1 | 22 | 3923 | |||
| NM_153202.3 | NP_694882.1 | 21 | 3845 | |||
| XM_005260843.1 | XP_005260900.1 | 22 | 3380 | |||
| XM_006723639.1 | XP_006723702.1 | 22 | 3377 | |||
| XM_006723640.1 | XP_006723703.1 | 22 | 3371 | |||
| XM_011529366.1 | XP_011527668.1 | 22 | 3279 | |||
| XM_011529367.1 | XP_011527669.1 | 22 | 3240 | |||
| XM_011529368.1 | XP_011527670.1 | 21 | 3302 | |||
| XM_011529373.1 | XP_011527675.1 | 21 | 3354 | |||
| GenBank mRNA | AF466287.1 | - | 22 | 3239 | ||
| AK092627.1 | - | 8 | 2117 | |||
| AK094070.1 | - | 2 | 2167 | |||
| AK095953.1 | - | 14 | 3025 | |||
| AK123015.1 | - | 18 | 3944 | |||
| AK126487.1 | - | 16 | 4921 | |||
| AK126498.1 | - | 10 | 4680 | |||
| AK226148.1 | - | 12 | 4984 | |||
| AL117415.1 | - | 11 | 2798 | |||
| AY358314.1 | - | 22 | 3351 | |||
| BX103461.1 | - | 1 | 535 | |||

ADAM33
Genomic Map