siRNA Details
| Gene Symbol: PRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077497.2 | NP_001070965.1 | 3 | 1734 | ||
| NM_025263.3 | NP_079539.2 | 4 | 1797 | |||
| GenBank mRNA | AK074531.1 | - | 3 | 1163 | ||
| AK131382.1 | - | 2 | 3125 | |||
| AK298621.1 | - | 4 | 1521 | |||
| U63336.1 | - | 4 | 1620 | |||

PRR3
Genomic Map