siRNA Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171170.1 | NP_001164641.1 | 5, 6 | 643 | ||
| NM_030631.3 | NP_085134.1 | 5, 6 | 643 | |||
| XM_011537287.2 | XP_011535589.1 | 6, 7 | 425 | |||
| XM_011537288.2 | XP_011535590.1 | 5, 6 | 517 | |||
| XM_011537289.2 | XP_011535591.1 | 4, 5 | 426 | |||
| GenBank mRNA | AJ278148.1 | - | 5, 6 | 643 | ||
| AK289575.1 | - | 5, 6 | 406 | |||
| BC101521.1 | - | 5, 6 | 379 | 8069027 | ||
| BC113365.1 | - | 5, 6 | 379 | 8322417 | ||

SLC25A21
Genomic Map