siRNA Details
| Gene Symbol: TMPRSS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288749.1 | NP_001275678.1 | 10 | 1326 | ||
| NM_001288750.1 | NP_001275679.1 | 12 | 1533 | |||
| NM_001288751.1 | NP_001275680.1 | 13 | 1740 | |||
| NM_001288752.1 | NP_001275681.1 | 11 | 1429 | |||
| NM_030770.3 | NP_110397.2 | 13 | 1636 | |||
| NR_110046.1 | 11 | 1434 | ||||
| NR_110047.1 | 10 | 1281 | ||||
| XM_017018366.1 | XP_016873855.1 | 10 | 6100 | |||
| XM_017018367.1 | XP_016873856.1 | 12 | 6292 | |||
| GenBank mRNA | AB028140.1 | - | 13 | 1642 | ||
| AK294313.1 | - | 10 | 1261 | |||
| AK295366.1 | - | 11 | 1358 | |||
| BC121802.1 | - | 12 | 1506 | 40085103 | ||
| BC121803.1 | - | 13 | 1713 | 40085104 | ||

TMPRSS5
Genomic Map