siRNA Details
| Gene Symbol: UCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003356.3 | NP_003347.1 | 4, 5 | 904 | ||
| NM_022803.2 | NP_073714.1 | 4, 5 | 904 | |||
| GenBank mRNA | AF001787.1 | - | 4, 5 | 732 | ||
| AF011449.1 | - | 3, 4 | 647 | |||
| AK092685.1 | - | 4, 5 | 551 | |||
| AK290630.1 | - | 4, 5 | 752 | |||
| AK291889.1 | - | 4, 5 | 718 | |||
| AK309873.1 | - | 4, 5 | 752 | |||
| KU178372.1 | - | 3, 4 | 549 | |||
| U82818.1 | - | 4, 5 | 702 | |||
| U84763.1 | - | 4, 5 | 702 | |||

UCP3
Genomic Map