siRNA Details
| Gene Symbol: PDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030815.2 | NP_110442.1 | 5 | 595 | ||
| GenBank mRNA | AY286301.1 | - | 5 | 607 | ||
| BC001856.2 | - | 5 | 466 | |||
| BC009334.2 | - | 5 | 520 | 4126826 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDRG1| Gene Symbol: PDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030815.2 | NP_110442.1 | 5 | 595 | ||
| GenBank mRNA | AY286301.1 | - | 5 | 607 | ||
| BC001856.2 | - | 5 | 466 | |||
| BC009334.2 | - | 5 | 520 | 4126826 | ||