siRNA Details
| Gene Symbol: CDADC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193478.1 | NP_001180407.1 | 3 | 337 | ||
| NM_030911.3 | NP_112173.1 | 3 | 337 | |||
| NR_036439.1 | 3 | 337 | ||||
| XM_011535250.2 | XP_011533552.1 | 3 | 301 | |||
| XM_011535251.1 | XP_011533553.1 | 3 | 281 | |||
| XM_011535253.1 | XP_011533555.1 | 3 | 373 | |||
| XM_017020778.1 | XP_016876267.1 | 1 | 20 | |||
| XM_017020779.1 | XP_016876268.1 | 3 | 329 | |||
| XM_017020780.1 | XP_016876269.1 | 3 | 679 | |||
| XM_017020781.1 | XP_016876270.1 | 3 | 329 | |||
| XM_017020782.1 | XP_016876271.1 | 2 | 127 | |||
| XM_017020783.1 | XP_016876272.1 | 3 | 679 | |||
| GenBank mRNA | AK297400.1 | - | 3 | 301 | ||
| AK312835.1 | - | 3 | 307 | |||
| AY027525.1 | - | 3 | 286 | |||
| BC009562.1 | - | 3 | 301 | |||
| BC032889.1 | - | 3 | 312 | |||
| BC048092.1 | - | 3 | 273 | |||
| BC070238.1 | - | 3 | 305 | |||
| BC092434.1 | - | 3 | 315 | |||
| BC125202.1 | - | 3 | 251 | 40123430 | ||
| DA400049.1 | - | 3 | 337 | |||
| DA560379.1 | - | 3 | 337 | |||

CDADC1
Genomic Map