siRNA Details
| Gene Symbol: URM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135947.2 | NP_001129419.1 | 4 | 1462 | ||
| NM_001265582.1 | NP_001252511.1 | 3 | 2825 | |||
| NM_030914.3 | NP_112176.1 | 5 | 1106 | |||
| NR_049743.1 | 5 | 1147 | ||||
| GenBank mRNA | AK001880.1 | - | 5 | 1055 | ||
| AK097029.1 | - | 3 | 2771 | |||
| BC003581.1 | - | 5 | 1064 | 3546655 | ||
| D83200.1 | - | 4 | 1009 | |||

URM1
Genomic Map