siRNA Details
| Gene Symbol: KAZALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030929.4 | NP_112191.2 | 5 | 2439 | ||
| GenBank mRNA | AK172862.1 | - | 5 | 1690 | ||
| BC007758.2 | - | 5 | 1840 | 4122366 | ||
| DA422973.1 | - | 1 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KAZALD1| Gene Symbol: KAZALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030929.4 | NP_112191.2 | 5 | 2439 | ||
| GenBank mRNA | AK172862.1 | - | 5 | 1690 | ||
| BC007758.2 | - | 5 | 1840 | 4122366 | ||
| DA422973.1 | - | 1 | 324 | |||