siRNA Details
| Gene Symbol: TIGD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243253.1 | NP_001230182.1 | 2 | 917 | ||
| NM_030953.3 | NP_112215.1 | 2 | 930 | |||
| GenBank mRNA | AK056604.1 | - | 2 | 418 | ||
| AK092978.1 | - | 2 | 917 | |||
| AK096325.1 | - | 2 | 930 | |||
| AL136539.1 | - | 2 | 341 | |||
| BC033641.1 | - | 2 | 878 | |||
| BC117249.1 | - | 1 | 231 | 40125800 | ||
| CR533559.1 | - | 1 | 155 | |||

TIGD6
Genomic Map