siRNA Details
Gene Symbol: OR12D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030959.2 | NP_112221.1 | 1 | 1182 | ||
GenBank mRNA | AJ459858.1 | - | 1 | 1182 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR12D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030959.2 | NP_112221.1 | 1 | 1182 | ||
GenBank mRNA | AJ459858.1 | - | 1 | 1182 |