siRNA Details
| Gene Symbol: TMEM14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127711.2 | NP_001121183.1 | 5 | 607 | ||
| NM_001286484.1 | NP_001273413.1 | 5 | 632 | |||
| NM_030969.4 | NP_112231.3 | 6 | 709 | |||
| NR_104454.1 | 4 | 530 | ||||
| GenBank mRNA | AK291883.1 | - | 6 | 663 | ||
| BC001033.1 | - | 6 | 652 | 3139565 | ||
| BC007080.1 | - | 6 | 664 | 4066217 | ||
| BC013913.1 | - | 6 | 671 | 3685635 | ||
| BC071660.1 | - | 6 | 653 | 4473732 | ||
| BU954633.1 | - | 4 | 472 | |||

TMEM14B
Genomic Map