siRNA Details
| Gene Symbol: PITPNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165966.1 | NP_001159438.1 | 14 | 1959 | ||
| NM_031220.3 | NP_112497.2 | 15 | 2067 | |||
| XM_011524015.2 | XP_011522317.1 | 15 | 2007 | |||
| XM_011524016.2 | XP_011522318.1 | 15 | 2008 | |||
| GenBank mRNA | AB209130.1 | - | 9 | 4430 | ||
| AF334586.1 | - | 15 | 2064 | |||
| AL389994.1 | - | 14 | 1790 | |||
| BC020484.1 | - | 6 | 665 | |||
| BC128583.1 | - | 15 | 2004 | 40128716 | ||
| BC128584.1 | - | 15 | 2004 | 40128724 | ||

PITPNM3
Genomic Map