siRNA Details
| Gene Symbol: RILP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031430.2 | NP_113618.2 | 8 | 1756 | ||
| XM_005256811.1 | XP_005256868.1 | 7 | 1651 | |||
| XM_017025195.1 | XP_016880684.1 | 6 | 1505 | |||
| GenBank mRNA | AF370391.1 | - | 8 | 1693 | ||
| AJ278711.1 | - | 8 | 1756 | |||
| AJ404317.1 | - | 8 | 1726 | |||
| AK055755.1 | - | 8 | 1735 | |||
| BC004961.1 | - | 8 | 1305 | |||
| BC031621.1 | - | 8 | 1550 | 5174974 | ||

RILP
Genomic Map