siRNA Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300741.1 | NP_001287670.1 | 2 | 201 | ||
| NM_031438.3 | NP_113626.1 | 2 | 201 | |||
| XM_005272095.1 | XP_005272152.1 | 2 | 186 | |||
| XM_005272097.3 | XP_005272154.1 | 2 | 201 | |||
| GenBank mRNA | AK098066.1 | - | 2 | 170 | ||
| AK304010.1 | - | 2 | 189 | |||
| AK309942.1 | - | 2 | 186 | |||
| AL136592.1 | - | 2 | 156 | |||
| BC041099.1 | - | 2 | 185 | 5495144 | ||
| CB131118.1 | - | 2 | 189 | |||
| DB029218.1 | - | 2 | 203 | |||
| DC417976.1 | - | 2 | 190 | |||

NUDT12
Genomic Map