siRNA Details
| Gene Symbol: FAM107B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282695.1 | NP_001269624.1 | 1 | 630 | ||
| NM_031453.3 | NP_113641.2 | 1 | 630 | |||
| GenBank mRNA | AK127413.1 | - | 1 | 630 | ||
| BC144707.1 | - | 1 | 438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM107B| Gene Symbol: FAM107B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282695.1 | NP_001269624.1 | 1 | 630 | ||
| NM_031453.3 | NP_113641.2 | 1 | 630 | |||
| GenBank mRNA | AK127413.1 | - | 1 | 630 | ||
| BC144707.1 | - | 1 | 438 | |||