siRNA Details
| Gene Symbol: RIOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031480.2 | NP_113668.2 | 13 | 1415 | ||
| NM_153005.1 | NP_694550.1 | 6 | 725 | |||
| XM_005249446.3 | XP_005249503.1 | 13 | 1460 | |||
| XM_011514933.2 | XP_011513235.2 | 13 | 1377 | |||
| GenBank mRNA | AF258661.1 | - | 6 | 725 | ||
| AK054568.1 | - | 13 | 1406 | |||
| AK314467.1 | - | 13 | 1444 | |||
| AL834277.1 | - | 12 | 1114 | |||
| BC006104.1 | - | 13 | 1415 | 3537086 | ||

RIOK1
Genomic Map