siRNA Details
| Gene Symbol: RSPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031924.4 | NP_114130.3 | 7 | 1532 | ||
| XM_005267153.4 | XP_005267210.1 | 5 | 1510 | |||
| XM_017011347.1 | XP_016866836.1 | 6 | 853 | |||
| GenBank mRNA | AF353618.1 | - | 7 | 1131 | ||
| AK057655.1 | - | 5 | 859 | |||
| AK057931.1 | - | 7 | 1553 | |||
| BC050604.1 | - | 7 | 1532 | 6161669 | ||

RSPH3
Genomic Map