siRNA Details
Gene Symbol: GPR61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031936.4 | NP_114142.3 | 2 | 2107 | ||
XM_017002438.1 | XP_016857927.1 | 2 | 4440 | |||
GenBank mRNA | AF258342.1 | - | 3 | 1743 | ||
AK290027.1 | - | 2 | 2107 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
