siRNA Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011544271.2 | XP_011542573.1 | 4 | 1795 | ||
| GenBank mRNA | AB060283.1 | - | 4 | 1235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WNT9A| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011544271.2 | XP_011542573.1 | 4 | 1795 | ||
| GenBank mRNA | AB060283.1 | - | 4 | 1235 | ||