siRNA Details
| Gene Symbol: MRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127174.1 | NP_001120646.1 | 6 | 1372 | ||
| NM_001127175.1 | NP_001120647.1 | 6 | 1483 | |||
| NM_001127176.1 | NP_001120648.1 | 7 | 1639 | |||
| NM_031939.3 | NP_114145.2 | 8 | 1742 | |||
| XM_005258365.3 | XP_005258422.1 | 7 | 1552 | |||
| XM_011526226.2 | XP_011524528.1 | 7 | 1523 | |||
| XM_017026035.1 | XP_016881524.1 | 7 | 1519 | |||
| XM_017026036.1 | XP_016881525.1 | 7 | 1523 | |||
| XM_017026037.1 | XP_016881526.1 | 6 | 1512 | |||
| GenBank mRNA | AK054702.1 | - | 8 | 1676 | ||
| AK295666.1 | - | 6 | 1484 | |||

MRO
Genomic Map