siRNA Details
| Gene Symbol: RNF26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032015.4 | NP_114404.1 | 1 | 1570 | ||
| GenBank mRNA | AB055622.1 | - | 1 | 1258 | ||
| AK075379.1 | - | 1 | 1328 | |||
| AK122642.1 | - | 1 | 1543 | |||
| BC000058.2 | - | 1 | 1339 | 3507662 | ||
| BC007534.1 | - | 1 | 1310 | 3028875 | ||
| BX647424.1 | - | 1 | 1543 | |||
| BX648447.1 | - | 1 | 1546 | |||

RNF26
Genomic Map