siRNA Details
| Gene Symbol: REG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159352.1 | NP_001152824.1 | 5 | 678 | ||
| NM_032044.3 | NP_114433.1 | 4 | 504 | |||
| GenBank mRNA | AF254415.1 | - | 4 | 419 | ||
| AF345934.1 | - | 4 | 372 | |||
| AY007243.1 | - | 4 | 360 | |||
| AY126670.1 | - | 5 | 678 | |||
| AY126671.1 | - | 4 | 504 | |||
| BC017089.2 | - | 4 | 376 | 3850358 | ||

REG4
Genomic Map