siRNA Details
| Gene Symbol: KBTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091344.1 | - | 1 | 1446 | ||
| AK299614.1 | - | 3 | 1080 | |||
| AK315589.1 | - | 1 | 1441 | |||
| AL136782.1 | - | 1 | 1358 | |||
| BC022033.2 | - | 1 | 1441 | 4825536 | ||
| CR533558.1 | - | 1 | 1132 | |||
| FJ150424.1 | - | 1 | 1441 | |||

KBTBD7
Genomic Map