siRNA Details
| Gene Symbol: C9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317997.1 | NP_001304926.1 | 5 | 1738 | ||
| NM_032307.4 | NP_115683.3 | 4 | 2136 | |||
| GenBank mRNA | AK090882.1 | - | 5 | 1741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf64| Gene Symbol: C9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317997.1 | NP_001304926.1 | 5 | 1738 | ||
| NM_032307.4 | NP_115683.3 | 4 | 2136 | |||
| GenBank mRNA | AK090882.1 | - | 5 | 1741 | ||