siRNA Details
| Gene Symbol: ING5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032329.4 | NP_115705.2 | 8 | 781 | ||
| XM_017005097.1 | XP_016860586.1 | 8 | 1849 | |||
| XM_017005098.1 | XP_016860587.1 | 9 | 1898 | |||
| GenBank mRNA | AF189286.1 | - | 8 | 758 | ||
| AK074422.1 | - | 8 | 781 | |||
| AK289958.1 | - | 8 | 782 | |||
| BC071899.1 | - | 8 | 772 | 6111003 | ||

ING5
Genomic Map