siRNA Details
| Gene Symbol: ACBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032360.3 | NP_115736.1 | 6 | 1236 | ||
| GenBank mRNA | AK314112.1 | - | 6 | 1211 | ||
| BC006505.2 | - | 6 | 693 | 2821736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACBD6| Gene Symbol: ACBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032360.3 | NP_115736.1 | 6 | 1236 | ||
| GenBank mRNA | AK314112.1 | - | 6 | 1211 | ||
| BC006505.2 | - | 6 | 693 | 2821736 | ||