siRNA Details
| Gene Symbol: ARFGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242832.1 | NP_001229761.1 | 2 | 393 | ||
| NM_032389.4 | NP_115765.2 | 2 | 393 | |||
| XM_005253166.1 | XP_005253223.1 | 2 | 771 | |||
| XM_005253167.1 | XP_005253224.1 | 2 | 771 | |||
| XM_005253168.2 | XP_005253225.1 | 2 | 771 | |||
| XM_006718346.1 | XP_006718409.1 | 2 | 771 | |||
| XM_011520405.1 | XP_011518707.1 | 2 | 771 | |||
| XM_017018413.1 | XP_016873902.1 | 2 | 771 | |||
| XM_017018414.1 | XP_016873903.1 | 2 | 147 | |||
| XM_017018415.1 | XP_016873904.1 | 2 | 141 | |||
| GenBank mRNA | AK098324.1 | - | 2 | 147 | ||
| AK300347.1 | - | 2 | 127 | |||
| AK301785.1 | - | 2 | 97 | |||
| AK304541.1 | - | 2 | 132 | |||
| AK315992.1 | - | 2 | 141 | |||
| AL834337.1 | - | 2 | 112 | |||
| BC030148.2 | - | 2 | 136 | 4558453 | ||
| BC036823.1 | - | 2 | 138 | |||
| BM827445.1 | - | 2 | 393 | |||

ARFGAP2
Genomic Map