siRNA Details
| Gene Symbol: FAXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032511.2 | NP_115900.1 | 6 | 1299 | ||
| XM_006715582.2 | XP_006715645.1 | 4 | 499 | |||
| XM_011536188.2 | XP_011534490.1 | 6 | 816 | |||
| GenBank mRNA | AK057793.1 | - | 5 | 717 | ||
| AK096480.1 | - | 6 | 1062 | |||
| BC004869.2 | - | 6 | 1299 | 3834272 | ||
| BC006515.2 | - | 3 | 230 | |||
| BC011583.1 | - | 6 | 1299 | 4577456 | ||
| BQ883579.1 | - | 5 | 741 | |||

FAXC
Genomic Map