siRNA Details
| Gene Symbol: ZNF347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 3842 | ||
| NM_001172675.1 | NP_001166146.1 | 5 | 3522 | |||
| NM_032584.2 | NP_115973.2 | 5 | 3519 | |||
| XM_005259335.4 | XP_005259392.1 | 5 | 3841 | |||
| XM_005259336.4 | XP_005259393.1 | 5 | 3631 | |||
| XM_017027384.1 | XP_016882873.1 | 5 | 3861 | |||
| XM_017027385.1 | XP_016882874.1 | 5 | 3545 | |||
| XM_017027386.1 | XP_016882875.1 | 4 | 3467 | |||
| GenBank mRNA | AL713691.1 | - | 1 | 2500 | ||
| AY029765.1 | - | 5 | 3481 | |||

ZNF347
Genomic Map