siRNA Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032588.3 | NP_115977.2 | 2 | 388 | ||
| XM_017002559.1 | XP_016858048.1 | 2 | 388 | |||
| GenBank mRNA | AF353673.1 | - | 2 | 367 | ||
| AJ276484.1 | - | 2 | 371 | |||
| AK056942.1 | - | 2 | 384 | |||
| AK297820.1 | - | 2 | 384 | |||
| BC080529.1 | - | 2 | 366 | 4862321 | ||
| BT007373.1 | - | 2 | 248 | |||
| DA900348.1 | - | 2 | 388 | |||

TRIM63
Genomic Map