siRNA Details
| Gene Symbol: KDM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005366.1 | NP_001005366.1 | 10 | 1479 | ||
| NM_032590.4 | NP_115979.3 | 10 | 1269 | |||
| XM_005253955.3 | XP_005254012.1 | 10 | 1479 | |||
| XM_005253956.3 | XP_005254013.1 | 10 | 1076 | |||
| XM_011538867.2 | XP_011537169.1 | 10 | 2522 | |||
| XM_011538868.2 | XP_011537170.1 | 10 | 2518 | |||
| XM_011538869.2 | XP_011537171.1 | 10 | 2520 | |||
| GenBank mRNA | AJ459424.1 | - | 10 | 1241 | ||
| AK127328.1 | - | 10 | 1479 | |||
| AL133572.1 | - | 6 | 782 | |||
| BC115379.1 | - | 9 | 1129 | |||
| BC115380.1 | - | 10 | 1208 | 40029132 | ||
| BX648928.1 | - | 8 | 1001 | |||

KDM2B
Genomic Map