siRNA Details
| Gene Symbol: RTN4IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032730.5 | NP_116119.2 | 1 | 580 | ||
| XM_017011375.1 | XP_016866864.1 | 1 | 580 | |||
| XM_017011376.1 | XP_016866865.1 | 1 | 580 | |||
| GenBank mRNA | AF336861.1 | - | 1 | 154 | ||
| AF439711.1 | - | 1 | 103 | |||
| AY063761.1 | - | 1 | 103 | |||
| BC006399.2 | - | 1 | 375 | 4309263 | ||
| BE894352.1 | - | 518 | ||||
| BQ936729.1 | - | 1 | 558 | |||

RTN4IP1
Genomic Map