siRNA Details
| Gene Symbol: ADGRG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308362.1 | NP_001295291.1 | 6 | 1102 | ||
| NM_032787.2 | NP_116176.2 | 12 | 1926 | |||
| XM_011513245.2 | XP_011511547.1 | 11 | 1801 | |||
| GenBank mRNA | AK027360.1 | - | 12 | 1914 | ||
| AK122804.1 | - | 1 | 918 | |||
| AK131069.1 | - | 5 | 785 | |||
| AK172769.1 | - | 8 | 1168 | |||
| AK172774.1 | - | 4 | 679 | |||
| AK295345.1 | - | 6 | 1044 | |||
| AY181246.1 | - | 12 | 1658 | |||
| BC113451.1 | - | 12 | 1692 | 8322503 | ||
| BC130388.1 | - | 12 | 1692 | 40146419 | ||
More Information
Set Membership: |
|

ADGRG7
Genomic Map