siRNA Details
| Gene Symbol: ADO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032804.5 | NP_116193.2 | 1 | 2014 | ||
| GenBank mRNA | AK027453.1 | - | 1 | 1609 | ||
| AK127694.1 | - | 1 | 2016 | |||
| BC018660.1 | - | 1 | 1918 | 4104391 | ||
| BC028589.2 | - | 1 | 1884 | 4838951 | ||
| BC067740.1 | - | 1 | 1822 | 5295674 | ||

ADO
Genomic Map